Learn More
TARGETMOL CHEMICALS INC Iso-Isomucronulatol 7-O-glucos
SDP
Catalog No. 502879172
Encompass_Preferred
Agent icon Sure, here is the string without special characters Also available in 1 mL 1 mg 10 mg 25 mg 50 mg 100 mg and bulk Please contact Fisher for quotes 72-Dihydroxy-34-dimethoxyisoflavan 7-O-beta-D-glucoside Isomucronulatol 7-O-glucoside is a biologically active isoflavone isolated from Astragalus membranaceus Purity 9974%
Catalog No. 50-287-9172 Supplier TARGETMOL CHEMICALS INC Supplier No. T38485MG
Add to Cart
Add to Cart
Agent icon Sure, here is the string without special characters Also available in 1 mL 1 mg 10 mg 25 mg 50 mg 100 mg and bulk Please contact Fisher for quotes 72-Dihydroxy-34-dimethoxyisoflavan 7-O-beta-D-glucoside Isomucronulatol 7-O-glucoside is a biologically active isoflavone isolated from Astragalus membranaceus Purity 9974%