Learn More
World Precision Instrument ISO-NOP Startup Kit
SDP
Catalog No. 502521254
Encompass
Hit the ground running and get the supplies you need for your ISO-NOP Nitric Oxide Sensor
This kit contains everything you need to begin working with the ISO-NOP sensor, and the kit is highly recommended for first-time ISO-NOP users. The kit includes additional membrane sleeves (5), electrolyte filling solution, a MicroFil electrolyte filling needle, a 1 mL syringe, sample vials with caps (2), acrylic stand assembly, potentiometer adjustment tool (aka tweaker), rubber stoppers (2), rubber stoppers with holes (2), 25g needles (2) and a 1/8" ID male luer fitting.
Catalog No. 50-252-1254 Supplier World Precision Instrument Supplier No. 5435
Hit the ground running and get the supplies you need for your ISO-NOP Nitric Oxide Sensor
This kit contains everything you need to begin working with the ISO-NOP sensor, and the kit is highly recommended for first-time ISO-NOP users. The kit includes additional membrane sleeves (5), electrolyte filling solution, a MicroFil electrolyte filling needle, a 1 mL syringe, sample vials with caps (2), acrylic stand assembly, potentiometer adjustment tool (aka tweaker), rubber stoppers (2), rubber stoppers with holes (2), 25g needles (2) and a 1/8" ID male luer fitting.