Learn More
World Precision Instrument ISO-NOPNM Nitric Oxide Sensor - 100nm
SDP
Catalog No. 502521157
Encompass
The ISO-NOPNM is based on a novel design in which an electrochemically "activated" composite graphite nanofiber is used as the NO-sensing element. The surface of the Nanosensor is then modified using a unique multi-layered NO-selective membrane. Figure at right illustrates the response of the ISO-NOPNM following successive additions of nanomolar concentrations of NO. The ultra-low noise of the ISO-NOPNM (0.5 pA) enables a detection limit of just 0.5 nM NO. The response time of ISO-NOPNM is less than 3 seconds.
Catalog No. 50-252-1157 Supplier World Precision Instrument Supplier No. ISONOPNM
The ISO-NOPNM is based on a novel design in which an electrochemically "activated" composite graphite nanofiber is used as the NO-sensing element. The surface of the Nanosensor is then modified using a unique multi-layered NO-selective membrane. Figure at right illustrates the response of the ISO-NOPNM following successive additions of nanomolar concentrations of NO. The ultra-low noise of the ISO-NOPNM (0.5 pA) enables a detection limit of just 0.5 nM NO. The response time of ISO-NOPNM is less than 3 seconds.