Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iso-Octane, for HPLC, GC, pesticide residue analysis and spectrophotometry, >99.0%, Solstice

Catalog No. 6000868 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
4 x 4 L
6 x 1 L
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-008-68 6 x 1 L Glass Bottle
60-008-70 4 x 4 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-008-68 Supplier Solstice Supplier No. 3621L
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 540-84-1
Molecular Formula C8H18
Molecular Weight (g/mol) 114.232
InChI Key NHTMVDHEPJAVLT-UHFFFAOYSA-N
Synonym isooctane, isobutyltrimethylmethane, pentane, 2,2,4-trimethyl, iso-octane, 2,4,4-trimethylpentane, unii-qab8f5669o, ch3 2chch2c ch3 3, 2, 2, 4-trimethylpentane, i-octane, isobutyltrimethylethane
PubChem CID 10907
ChEBI CHEBI:62805
IUPAC Name 2,2,4-trimethylpentane
SMILES CC(C)CC(C)(C)C

Specifications

Absorbance 0.1 AU max. at 225 nm, 0.02 AU max. at 250 nm, 0.005 AU max. at 300 nm
CAS Max % 100.0000%
Density 0.69 g/cm3
Quantity 6 x 1 L
Flash Point −8°C
Packaging Glass Bottle
UN Number UN1262
Boiling Point 99.24°C
Formula Weight 114.23
Percent Purity >99%
Grade HPLC
Chemical Name or Material Iso-Octane, B&J Brand™, for HPLC, GC, pesticide residue analysis and spectrophotometry, >99.0%
Show More Show Less
ShelfLife 547 days from date of manufacture
DOTInformation Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Octanes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.