Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Jade Scientific, Inc ISO-OCTANE LABORATORY PLUS 19L
SDP

Catalog No. 50326316 Shop All Jade Scientific, Inc Products
Encompass_Preferred

Iso-octane Laboratory Plus 19l. 540-84-1 MFCD00008943

Catalog No. 50-326-316 Supplier Jade Scientific, Inc Supplier No. BJLP362D19
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Iso-octane Laboratory Plus 19l. 540-84-1 MFCD00008943

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.