Learn More
Cayman Chemical Iso-OlomoucIn 10mg
SDP
Catalog No. 502951866
Encompass_Preferred
An inactive stereoisomer (IC50 ≥ 1 mM) of the Cdk5 inhibitor olomoucine; may have utility as a control compound for determining Cdk5 specificity; rapidly inhibits dopamine transporter activity in rat dorsal striatal synaptosomes with an IC50 value of ~37 μM
Catalog No. 50-295-1866 Supplier Cayman Chemical Supplier No. 1332510 MG
An inactive stereoisomer (IC50 ≥ 1 mM) of the Cdk5 inhibitor olomoucine; may have utility as a control compound for determining Cdk5 specificity; rapidly inhibits dopamine transporter activity in rat dorsal striatal synaptosomes with an IC50 value of ~37 μM