Learn More
World Precision Instrument ISO-OXY-2 or OXELP Filling Solution
SDP
Catalog No. 502521235
Encompass
  • To use with ISO-OXY-2 or OXELP Oxygen Sensor
  • 10ml
Catalog No. 50-252-1235 Supplier World Precision Instrument Supplier No. 7326
  • To use with ISO-OXY-2 or OXELP Oxygen Sensor
  • 10ml