Learn More
IDEX Health & Science Upchurch Iso-Prep Filter, SS, 2 um, 21.2 mm
Supplier:  IDEX Health & Science Upchurch 703120
Encompass_Preferred
Protect larger columns and injections from harmful particulates. Ideal for higher-flow systems. Interchangeable frits make protection more economical. Compatible with SFC and HPLC. Available to fit a variety of tubing sizes.
Catalog No. NC1398087