Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iso-Propyl Alcohol, 70% v/v, Reagents

Catalog No. 01334904
Change view
Click to view available options
Quantity:
500 mL
4 L
20 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-334-904 4 L
01-334-905 20 L
01-334-903 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01-334-904 Supplier Reagents Holdings Llc Supplier No. CI2960004A
Add to Cart
Add to Cart

Iso-Propyl Alcohol, 70% v/v, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 67-63-0 , 7732-18-5
Molecular Formula C3H8O
Molecular Weight (g/mol) 60.10
InChI Key KFZMGEQAYNKOFK-UHFFFAOYSA-N
Synonym Isopropyl Alcohol, 2-Propanol, Propan-2-ol
IUPAC Name propan-2-ol
SMILES CC(C)O

Specifications

Color Colorless
Quantity 4 L
Packaging Poly Bottle
Physical Form Liquid
Specific Gravity 0.877
Boiling Point 88°C
Percent Purity 70%
Grade Reagent
Chemical Name or Material Iso-Propyl Alcohol, 70% v/v
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.