Learn More
Medchemexpress LLC Iso-VQA-ACC acetate | 99.48% | 625.63 (free base) | 1 MG
SDP
Catalog No. 5000424888
Encompass_Preferred
iso-VQA-ACC acetate is a substrate for constitutive proteasome, characterized by its molecular formula C29H35N7O9.xC2H4O2 and a molecular weight of 625.63 (free base). This compound has a purity of 99.48% and is commonly identified by its sequence, iso-Val-Gln-Ala-{ACC}. It is primarily used in research involving proteasome and metabolic enzyme/protease pathways.
  • Targets proteasome activity for metabolic studies.
  • High purity suitable for sensitive research applications.
  • Soluble in DMSO at 100 mg/mL, requiring ultrasonic treatment for dissolution.
Catalog No. 50-004-24888 Supplier Medchemexpress LLC Supplier No. HYP10003B1MG
iso-VQA-ACC acetate is a substrate for constitutive proteasome, characterized by its molecular formula C29H35N7O9.xC2H4O2 and a molecular weight of 625.63 (free base). This compound has a purity of 99.48% and is commonly identified by its sequence, iso-Val-Gln-Ala-{ACC}. It is primarily used in research involving proteasome and metabolic enzyme/protease pathways.
  • Targets proteasome activity for metabolic studies.
  • High purity suitable for sensitive research applications.
  • Soluble in DMSO at 100 mg/mL, requiring ultrasonic treatment for dissolution.