Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Iso-VQA-ACC (acetate) | 99.48% | 625.63 (free base) | 10 MG
SDP

Catalog No. 5000424960
Encompass_Preferred

Iso-VQA-ACC acetate is a substrate for constitutive proteasome, designed for research applications. This compound is provided as a white to off-white solid with a high purity of 99.48% and a molecular weight of 625.63 (free base).

  • Substrate for constitutive proteasome
  • Molecular formula: C29H35N7O9.xC2H4O2
  • Sequence: iso-Val-Gln-Ala-{ACC}
  • Target: Proteasome
  • Pathway: Metabolic Enzyme/Protease
  • Powder storage: -80°C for 2 years, -20°C for 1 year (sealed, away from moisture)
  • In solvent storage: -80°C for 6 months, -20°C for 1 month (sealed, away from moisture)
  • Solubility: 100 mg/mL in DMSO (ultrasonic assistance)
  • Appearance: White to off-white solid

Catalog No. 50-004-24960 Supplier Medchemexpress LLC Supplier No. HYP10003B10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Iso-VQA-ACC acetate is a substrate for constitutive proteasome, designed for research applications. This compound is provided as a white to off-white solid with a high purity of 99.48% and a molecular weight of 625.63 (free base).

  • Substrate for constitutive proteasome
  • Molecular formula: C29H35N7O9.xC2H4O2
  • Sequence: iso-Val-Gln-Ala-{ACC}
  • Target: Proteasome
  • Pathway: Metabolic Enzyme/Protease
  • Powder storage: -80°C for 2 years, -20°C for 1 year (sealed, away from moisture)
  • In solvent storage: -80°C for 6 months, -20°C for 1 month (sealed, away from moisture)
  • Solubility: 100 mg/mL in DMSO (ultrasonic assistance)
  • Appearance: White to off-white solid

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.