Learn More
Medchemexpress LLC Iso-VQA-ACC acetate | 99.5% | 625.63 | 5 MG
SDP
Catalog No. 5000424961
Encompass_Preferred
Iso-VQA-ACC acetate is a substrate for constitutive proteasome, typically appearing as a white to off-white solid.
  • Functions as a substrate for constitutive proteasome
  • Targets proteasome activity
  • Involved in the metabolic enzyme/protease pathway
  • Molecular sequence: iso-Val-Gln-Ala-{ACC}
  • Achieves a purity of 99.5% by HPLC
  • Structure confirmed by MS analysis
Catalog No. 50-004-24961 Supplier Medchemexpress LLC Supplier No. HYP10003B5MG
Iso-VQA-ACC acetate is a substrate for constitutive proteasome, typically appearing as a white to off-white solid.
  • Functions as a substrate for constitutive proteasome
  • Targets proteasome activity
  • Involved in the metabolic enzyme/protease pathway
  • Molecular sequence: iso-Val-Gln-Ala-{ACC}
  • Achieves a purity of 99.5% by HPLC
  • Structure confirmed by MS analysis