Learn More
GA International ISO Warning Labels for Deep-Freeze Applications - "Low Temperature Symbol" 0.75" x 0.75" / 19.1mm x 19.1mm Rectangle Black & Yellow on Silver From -80°C to +100°C (-112°F to +212°F)
Catalog No. 5000571874
Encompass_Preferred
Deep-freeze pre-printed warning labels for the clear identification of low temperature hazards as well as containers and samples that need to be stored in low temperature conditions. Provided with a glove-friendly adhesive, these waterproof freezer labels are perfect for labeling microtubes, vials, as well as cardboard and polypropylene freezer boxes for storage in laboratory freezers (-80°C/-112°F; -40°C/-40°F; -20°C/-4°F) or transported on dry ice. Labels can be removed at room temperature without leaving behind any adhesive residue.
Catalog No. 50-005-71874 Supplier GA International Supplier No. WL0100.25R
Deep-freeze pre-printed warning labels for the clear identification of low temperature hazards as well as containers and samples that need to be stored in low temperature conditions. Provided with a glove-friendly adhesive, these waterproof freezer labels are perfect for labeling microtubes, vials, as well as cardboard and polypropylene freezer boxes for storage in laboratory freezers (-80°C/-112°F; -40°C/-40°F; -20°C/-4°F) or transported on dry ice. Labels can be removed at room temperature without leaving behind any adhesive residue.