Learn More
GA International ISO Warning Labels - "General Warning Symbol" 0.75" x 0.75" / 19.1mm x 19.1mm Rectangle Black & Yellow on Silver From -80°C to +100°C (-112°F to +212°F)
Catalog No. 5000571872
Encompass_Preferred
Pre-printed general warning labels for identifying equipment, containers, and material that may pose a safety risk in the laboratory environment. Provided with a glove-friendly adhesive, these waterproof general use labels can be applied on plastic, glass, cardboard, and metal surfaces. With a wide temperature resistance range, the labels can be used in lab freezers and refrigerators, at room temperature (doors, drawers, etc.), hot surfaces, including water baths and heating blocks, as well as on equipment, instruments, and tools with moving parts.
Catalog No. 50-005-71872 Supplier GA International Supplier No. WL0090.25R
Pre-printed general warning labels for identifying equipment, containers, and material that may pose a safety risk in the laboratory environment. Provided with a glove-friendly adhesive, these waterproof general use labels can be applied on plastic, glass, cardboard, and metal surfaces. With a wide temperature resistance range, the labels can be used in lab freezers and refrigerators, at room temperature (doors, drawers, etc.), hot surfaces, including water baths and heating blocks, as well as on equipment, instruments, and tools with moving parts.