Learn More
Medchemexpress LLC Isoallolithocholic acid (3β-hydroxy-5α-cholanic acid) | 2276-93-9 | 99.8% | 250 MG
SDP
Catalog No. 5000405809
Encompass_Preferred
Isoallolithocholic acid, also known as 3β-Hydroxy-5α-cholanic acid, is a derivative of Lithocholic acid. It functions as a T cell regulator, enhancing the differentiation of regulatory T cells (Tregs). This product is intended for research use only.
  • Enhances regulatory T cells (Tregs) differentiation
  • Reduces Th17 cell differentiation by approximately 50%
  • Does not affect T cell differentiation into Th1 or Th2 cells
  • Enhanced expression of FoxP3 requires mitoROS
  • Appears as a white to off-white solid
Catalog No. 50-004-05809 Supplier Medchemexpress LLC Supplier No. 250MGHYB0172A
Add to Cart
Add to Cart
Isoallolithocholic acid, also known as 3β-Hydroxy-5α-cholanic acid, is a derivative of Lithocholic acid. It functions as a T cell regulator, enhancing the differentiation of regulatory T cells (Tregs). This product is intended for research use only.
  • Enhances regulatory T cells (Tregs) differentiation
  • Reduces Th17 cell differentiation by approximately 50%
  • Does not affect T cell differentiation into Th1 or Th2 cells
  • Enhanced expression of FoxP3 requires mitoROS
  • Appears as a white to off-white solid