Learn More
Medchemexpress LLC Isoallolithocholic acid-d2 | 2410277-69-7 | 99.3% | 1 MG
SDP
Catalog No. 5000405996
Encompass_Preferred
Isoallolithocholic acid-d2 is a deuterium-labeled compound, functioning as a T cell regulator by enhancing the differentiation of regulatory T cells (Tregs). Deuterium-labeled compounds are often used as tracers for quantification during drug development and can influence pharmacokinetic and metabolic profiles. This compound can also serve as an internal standard for quantitative analysis.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-05996 Supplier Medchemexpress LLC Supplier No. HYB0172AS1MG
Isoallolithocholic acid-d2 is a deuterium-labeled compound, functioning as a T cell regulator by enhancing the differentiation of regulatory T cells (Tregs). Deuterium-labeled compounds are often used as tracers for quantification during drug development and can influence pharmacokinetic and metabolic profiles. This compound can also serve as an internal standard for quantitative analysis.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS