Learn More
Cayman Chemical Isoalantolactone, Purity: ≥98%, CAS: 470-17-7, 250 mg
SDP
Catalog No. 502949493
Encompass_Preferred
A sesquiterpene lactone with diverse biological activities; inhibits proliferation of PANC-1, BxPC-3, and HPAC pancreatic cancer cells (IC50s = 40, 43, and 48 µM, respectively); induces apoptosis, cell cycle arrest at the S phase, and the production of ROS in PANC-1 cells; decreases LPS-induced production of NO, PGE2, IL-6, and TNF-α in RAW 264.7 macrophages at 4 and 8 µM; reduces production of S. aureus α-toxin in a concentration-dependent manner and increases survival in a mouse model of S. aureus-induced pneumonia at 50 mg/kg
Catalog No. 50-294-9493 Supplier Cayman Chemical Supplier No. 33838250 MG
A sesquiterpene lactone with diverse biological activities; inhibits proliferation of PANC-1, BxPC-3, and HPAC pancreatic cancer cells (IC50s = 40, 43, and 48 µM, respectively); induces apoptosis, cell cycle arrest at the S phase, and the production of ROS in PANC-1 cells; decreases LPS-induced production of NO, PGE2, IL-6, and TNF-α in RAW 264.7 macrophages at 4 and 8 µM; reduces production of S. aureus α-toxin in a concentration-dependent manner and increases survival in a mouse model of S. aureus-induced pneumonia at 50 mg/kg