Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isoamyl 4-methoxycinnamate, ≥98.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111015333 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-5333 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-5333 Supplier MilliporeSigma Supelco Supplier No. 4418950MG
Add to Cart
Add to Cart

Isoamyl 4-methoxycinnamate is a derivative of cinnamon acid ester and may be used as a sunscreen agent since it acts as a waterproof filter for ultraviolet B (UVB) rays.

Cleaning Products, Cosmetics, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

CAS 71617-10-2
Physical Form Neat
Linear Formula C15H20O3
MDL Number MFCD00583856
Synonym Isoamyl p-methoxycinnamate; 4-Methoxycinnamic acid 3-methylbutyl ester; 4-Methoxycinnamic acid isoamyl ester; Amiloxate; Isopentyl 4-methoxycinnamate; NSC 408332
Type Analytical Standard
Quantity 50 mg
Formula Weight 248.32
Percent Purity ≥98.0% (HPLC)
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.