Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isoamyl acetate, 99%, pure, mixture of 2- and 3-Methylbutyl acetate

Catalog No. AC150660025
Change view
Click to view available options
Quantity:
250 mL
1 L
2.5 L
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC150660025 2.5 L Glass bottle
AC150662500 250 mL Glass bottle
AC150660010 1 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. AC150660025 Supplier Thermo Scientific Chemicals Supplier No. 150660025
Only null left
Add to Cart
This item is not returnable. View return policy

CAS: 123-92-2 | C7H14O2 | 130.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123-92-2
Molecular Formula C7H14O2
Molecular Weight (g/mol) 130.19
MDL Number MFCD00008946
InChI Key MLFHJEHSLIIPHL-UHFFFAOYSA-N
Synonym isoamyl acetate, isopentyl acetate, isopentyl ethanoate, banana oil, pear oil, 3-methylbutyl ethanoate, isoamyl ethanoate, iso-amyl acetate, 3-methyl-1-butyl acetate, i-amyl acetate
PubChem CID 31276
ChEBI CHEBI:31725
IUPAC Name 3-methylbutyl acetate
SMILES CC(C)CCOC(C)=O

Specifications

Melting Point -78.0°C
Density 0.8740g/mL
Boiling Point 142.0°C (756.0 mmHg)
Flash Point 25°C
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Refractive Index 1.3990 to 1.4010
Linear Formula CH3CO2CH2CH2CH(CH3)2
Quantity 2.5 L
Beilstein 02, 132
Specific Gravity 0.874
Merck Index 15, 5156
Solubility Information Solubility in water: 2mg/mL (20°C). Other solubilities: soluble in ethanol,diethyl phthalate,mineral oil,and 1,2-propanediol
Viscosity 1.03 Pas (8.97°C)
Formula Weight 130.19
Percent Purity 99+%
Grade Pure
Chemical Name or Material Isoamyl acetate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Flammable liquid and vapor.
Repeated exposure may cause skin dryness or cracking.
Precautionary Statement GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 204-662-3

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.