Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isoamyl Acetoacetate 97.0+%, TCI America™
SDP

Catalog No. A081225ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A081225ML 25 mL
A0812500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A081225ML Supplier TCI America Supplier No. A081225ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2308-18-1
Molecular Formula C9H16O3
Molecular Weight (g/mol) 172.224
MDL Number MFCD00059355
InChI Key XHRGPLDMNNGHCX-UHFFFAOYSA-N
Synonym Acetoacetic Acid Isoamyl Ester, Isopentyl Acetoacetate
PubChem CID 61296
IUPAC Name 3-methylbutyl 3-oxobutanoate
SMILES CC(C)CCOC(=O)CC(=O)C

Specifications

Color Colorless
Boiling Point 218°C
Quantity 25 mL
Formula Weight 172.22
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Isoamyl Acetoacetate
EINECSNumber (2)-1505
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.