Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isoamyl Alcohol (Molecular Biology), Fisher BioReagents

Catalog No. BP1150500 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
500 mL
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
BP1150-500 500 mL Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BP1150-500

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 123-51-3 | C5H12O | 88.15 g/mol

500mL; Glass Bottle

A mixture of Chloroform and Isoamyl Alcohol (24:1 v/v) is used to remove proteins during nucleic acid extraction procedures. It may also be used to extract Ethidium Bromide from nucleic acids.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Isoamyl Alcohol
Acidity 0.01% max.
CAS 123-51-3
Melting Point -117°C
pH 6.5
Boiling Point 130°C
Assay Percent Range ≥99 %
Packaging Glass Bottle
Molecular Formula C5H12O
Linear Formula (CH3)2CHCH2CH2OH
Quantity 500 mL
Residue after Evaporation 0.003% max.
Merck Index 15, 5241
Synonym 3-methyl-1-butanol, isoamyl alcohol, isopentyl alcohol, isopentanol, 3-methylbutanol, 1-butanol, 3-methyl, isoamylol, isobutylcarbinol, iso-amylalkohol, isobutyl carbinol
InChI Key PHTQWCKDNZKARW-UHFFFAOYSA-N
SMILES CC(C)CCO
IUPAC Name 3-methylbutan-1-ol
Molecular Weight (g/mol) 88.15
PubChem CID 31260
ChEBI CHEBI:15837
Vapor Pressure 4hPa at 20°C
Formula Weight 88.15
Percent Purity ≥99%
Grade Molecular Biology
Physical Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.