Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isoamyl Benzoate 98.0+%, TCI America™
SDP

Catalog No. B007125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B007125G 25 g
B0071500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B007125G Supplier TCI America Supplier No. B007125G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 94-46-2
Molecular Formula C12H16O2
Molecular Weight (g/mol) 192.258
MDL Number MFCD00026515
InChI Key MLLAPOCBLWUFAP-UHFFFAOYSA-N
Synonym Benzoic Acid Isoamyl Ester, Isopentyl Benzoate
PubChem CID 7193
IUPAC Name 3-methylbutyl benzoate
SMILES CC(C)CCOC(=O)C1=CC=CC=C1

Specifications

Color Colorless
Boiling Point 262°C
Quantity 25 g
Formula Weight 192.26
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Isoamyl Benzoate
EINECSNumber (3)-1356
RTECSNumber DH3078000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.