Learn More
Medchemexpress LLC Isoamyl caprylate (isoamyl octanoate) | 2035-99-6 | MFCD00048917 | 99.4% | 214.35 | C13H26O2 | 50 G
SDP
Catalog No. 5000302098
Encompass_Preferred
Isopentyl octanoate is an ester (C13H26O2; MW 214.35; CAS 2035-99-6) formed from isoamyl alcohol and octanoic acid. It is used as a flavor and fragrance ingredient and as a reagent or analytical standard in biochemical applications.
  • Used as a flavor and fragrance ingredient in formulations.
  • Serves as a reagent or analytical standard in biochemical assays.
  • Has a fruity, pineapple-coconut aroma useful for scent profiles.
  • Offered in high purity suitable for analytical use (manufacturer lists 99.39%).
  • Molecular formula C13H26O2; molecular weight 214.35.
Catalog No. 50-003-02098 Supplier Medchemexpress LLC Supplier No. HYW01420650G
Isopentyl octanoate is an ester (C13H26O2; MW 214.35; CAS 2035-99-6) formed from isoamyl alcohol and octanoic acid. It is used as a flavor and fragrance ingredient and as a reagent or analytical standard in biochemical applications.
  • Used as a flavor and fragrance ingredient in formulations.
  • Serves as a reagent or analytical standard in biochemical assays.
  • Has a fruity, pineapple-coconut aroma useful for scent profiles.
  • Offered in high purity suitable for analytical use (manufacturer lists 99.39%).
  • Molecular formula C13H26O2; molecular weight 214.35.