Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isoamyl Decanoate 97.0+%, TCI America™
SDP

Catalog No. D001925ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D001925ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D001925ML Supplier TCI America Supplier No. D001925ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2306-91-4
Molecular Formula C15H30O2
Molecular Weight (g/mol) 242.403
MDL Number MFCD00053822
InChI Key XDOGFYDZGUDBQY-UHFFFAOYSA-N
Synonym Decanoic Acid Isoamyl Ester, Isopentyl Decanoate
PubChem CID 75320
IUPAC Name 3-methylbutyl decanoate
SMILES CCCCCCCCCC(=O)OCCC(C)C

Specifications

Color Colorless
Boiling Point 124°C
Quantity 25 mL
Formula Weight 242.40
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Isoamyl Decanoate
EINECSNumber (2)-0782&(2)-0798
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.