Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Isoamyl ether

Catalog No. 260649500ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
26-064-9500ML 500 mL
26-064-9100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 26-064-9500ML Supplier Sigma Aldrich Supplier No. 260649500ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Isoamyl ether has been used in the synthesis of:
  • Silver nanoparticles in a continuous flow tubular microreactor
  • Single-crystal ag nanocubes (with slight truncation) of around 13.5nm in edge length

Specifications

CAS 544-01-4
Density 0.778 g/mL (at 25°C (literature))
Boiling Point 173°C (lit.)
Molecular Formula C10H22O
Linear Formula (C5H11)2 O
Refractive Index n20/D 1.408 (literature)
MDL Number MFCD00008947
Quantity 500 mL
Synonym Isopentyl ether
Molecular Weight (g/mol) 158.28
Percent Purity 99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.