Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isoamyl Gallate 98.0+%, TCI America™
SDP

Catalog No. G001225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G001225G 25 g
G0012500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. G001225G Supplier TCI America Supplier No. G001225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2486-2-4
Molecular Formula C12H16O5
Molecular Weight (g/mol) 240.26
MDL Number MFCD00016431
InChI Key YBMTWYWCLVMFFD-UHFFFAOYSA-N
Synonym Gallic Acid Isoamyl Ester
PubChem CID 75596
IUPAC Name 3-methylbutyl 3,4,5-trihydroxybenzoate
SMILES CC(C)CCOC(=O)C1=CC(=C(C(=C1)O)O)O

Specifications

Melting Point 147°C
Color White-Yellow
Quantity 25 g
Formula Weight 240.26
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Isoamyl Gallate
EINECSNumber (3)-1583
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.