Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isoamyl Hexanoate (contains 2-Methylbutyl Hexanoate) 97.0+%, TCI America™
SDP

Catalog No. H010925ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H010925ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H010925ML Supplier TCI America Supplier No. H010925ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2198-61-0
Molecular Formula C11H22O2
Molecular Weight (g/mol) 186.295
MDL Number MFCD00027280
InChI Key XVSZRAWFCDHCBP-UHFFFAOYSA-N
Synonym Hexanoic Acid Isopentyl Ester, Isopentyl Hexanoate, Hexanoic Acid Isoamyl Ester
PubChem CID 16617
ChEBI CHEBI:87542
IUPAC Name 3-methylbutyl hexanoate
SMILES CCCCCC(=O)OCCC(C)C

Specifications

Color Yellow
Boiling Point 98°C
Quantity 25 mL
Formula Weight 186.30
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Isoamyl Hexanoate (contains 2-Methylbutyl Hexanoate)
EINECSNumber (2)-0782
RTECSNumber MO8389300
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.