Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isoamyl Hexanoate 98.0+%, TCI America™
SDP

Catalog No. I089925ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I089925ML 25 mL
I0899100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I089925ML Supplier TCI America Supplier No. I089925ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2198-61-0
Molecular Formula C11H22O2
Molecular Weight (g/mol) 186.295
MDL Number MFCD00027280
InChI Key XVSZRAWFCDHCBP-UHFFFAOYSA-N
Synonym Hexanoic Acid Isoamyl Ester, Isopentyl Hexanoate, Hexanoic Acid Isopentyl Ester
PubChem CID 16617
ChEBI CHEBI:87542
IUPAC Name 3-methylbutyl hexanoate
SMILES CCCCCC(=O)OCCC(C)C

Specifications

Color Colorless
Boiling Point 98°C
Quantity 25 mL
Formula Weight 186.30
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Isoamyl Hexanoate
EINECSNumber (2)-0782
RTECSNumber MO8389300
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.