Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isoamyl Propionate, MP Biomedicals

Catalog No. ICN210582 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN210582 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN210582 Supplier MP Biomedicals, Inc Supplier No. 0521058280
Add to Cart
Add to Cart

Chemical Identifiers

CAS 105-68-0
Molecular Formula C8H16O2
Molecular Weight (g/mol) 144.214
InChI Key XAOGXQMKWQFZEM-UHFFFAOYSA-N
Synonym isoamyl propionate, isoamyl propanoate, isopentyl propanoate, 3-methylbutyl propionate, isopentyl propionate, isopentyl alcohol, propionate, 1-butanol, 3-methyl-, propanoate, iso-pentyl propionate, propionic acid, isopentyl ester, 3-methyl-1-butyl propanoate
PubChem CID 7772
ChEBI CHEBI:87419
IUPAC Name 3-methylbutyl propanoate
SMILES CCC(=O)OCCC(C)C

Specifications

Quantity 100 g
Formula Weight 144.12
Chemical Name or Material Isoamyl Propionate
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.