Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isoamyl Salicylate (contains 2-Methylbutyl Salicylate), TCI America™
SDP

Catalog No. S001225ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S001225ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S001225ML Supplier TCI America Supplier No. S001225ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 87-20-7
Molecular Formula C12H16O3
Molecular Weight (g/mol) 208.257
MDL Number MFCD00020037
InChI Key PMGCQNGBLMMXEW-UHFFFAOYSA-N
Synonym Isopentyl Salicylate, Salicylic Acid Isoamyl Ester, Isopentyl 2-Hydroxybenzoate
PubChem CID 6874
IUPAC Name 3-methylbutyl 2-hydroxybenzoate
SMILES CC(C)CCOC(=O)C1=CC=CC=C1O

Specifications

Color Colorless
Boiling Point 273°C
Quantity 25 mL
UN Number 3082
Formula Weight 208.26
Physical Form Liquid
Chemical Name or Material Isoamyl Salicylate (contains 2-Methylbutyl Salicylate)
EINECSNumber (3)-1585
RTECSNumber VO4375000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.