Learn More
Medchemexpress LLC Isoangustone A | 129280-34-8 | 1 MILLIGRAM
SDP
Catalog No. 5000418870
Encompass_Preferred
Isoangustone A is an anticancer and anti-inflammatory agent. Isoangustone A induces cancer cells apoptosis and autophagic cell death.
  • Suppresses proliferation and induces G1 phase cell cycle arrest in SK-MEL-28 cells.
  • Decreases the abundance of G1 phase-related proteins mediated through Akt/GSK3β and MKK4/MKK7/JNKs signaling pathways.
  • Suppresses PI3-K, MKK4, and MKK7 kinase activities by directly binding in an ATP-competitive manner.
  • Induces autophagy in colorectal cancer cells by activating AMPK signaling.
  • Inhibits mitochondrial respiration.
  • Induces SW480 cells apoptosis.
  • Suppresses mesangial fibrosis and inflammation in human renal mesangial cells.
Catalog No. 50-004-18870 Supplier Medchemexpress LLC Supplier No. HYN40061MG
Isoangustone A is an anticancer and anti-inflammatory agent. Isoangustone A induces cancer cells apoptosis and autophagic cell death.
  • Suppresses proliferation and induces G1 phase cell cycle arrest in SK-MEL-28 cells.
  • Decreases the abundance of G1 phase-related proteins mediated through Akt/GSK3β and MKK4/MKK7/JNKs signaling pathways.
  • Suppresses PI3-K, MKK4, and MKK7 kinase activities by directly binding in an ATP-competitive manner.
  • Induces autophagy in colorectal cancer cells by activating AMPK signaling.
  • Inhibits mitochondrial respiration.
  • Induces SW480 cells apoptosis.
  • Suppresses mesangial fibrosis and inflammation in human renal mesangial cells.