Learn More
Medchemexpress LLC Isoasatone A | 67451-73-4 | 1 MG
SDP
Catalog No. 5000417895
Encompass_Preferred
Isoasatone A is a natural product derived from the plant Heterotropa takaoi M. It is recognized for its anti-insect activity, specifically targeting S. litura by influencing cytochrome P450 monoxygenases and glutathione transferases. This compound is exclusively for research purposes.
  • Isolated from Heterotropa takaoi M.
  • Exhibits anti-insect activity
  • Acts on cytochrome P450 monoxygenases and glutathione transferases
  • For research use only
Catalog No. 50-004-17895 Supplier Medchemexpress LLC Supplier No. HYN69941MG
Isoasatone A is a natural product derived from the plant Heterotropa takaoi M. It is recognized for its anti-insect activity, specifically targeting S. litura by influencing cytochrome P450 monoxygenases and glutathione transferases. This compound is exclusively for research purposes.
  • Isolated from Heterotropa takaoi M.
  • Exhibits anti-insect activity
  • Acts on cytochrome P450 monoxygenases and glutathione transferases
  • For research use only