Learn More
Cayman Chemical IsoastragalosIde I 10mg
SDP
Supplier:  Cayman Chemical 4027910 MG
Encompass_Preferred
A saponin with diverse biological activities; inhibits LPS-induced production of NO, TNF-α, and IL-1β, as well as inhibits NF-κB phosphorylation in BV-2 microglia from 25-100 µM; induces adiponectin production in 3T3-L1 adipocytes from 2-10 UG/ml; induces the expression of β-cell differentiation genes and increases mRNA levels of Ngn3 and the β-cell markers insulin1 and insulin2 in mPDOs
Catalog No. 50-294-3368