Learn More
Cayman Chemical IsoastragalosIde I 1mg
SDP
Catalog No. 502945887
Encompass_Preferred
A saponin with diverse biological activities; inhibits LPS-induced production of NO, TNF-α, and IL-1β, as well as inhibits NF-κB phosphorylation in BV-2 microglia from 25-100 µM; induces adiponectin production in 3T3-L1 adipocytes from 2-10 UG/ml; induces the expression of β-cell differentiation genes and increases mRNA levels of Ngn3 and the β-cell markers insulin1 and insulin2 in mPDOs
Catalog No. 50-294-5887 Supplier Cayman Chemical Supplier No. 402791 MG
A saponin with diverse biological activities; inhibits LPS-induced production of NO, TNF-α, and IL-1β, as well as inhibits NF-κB phosphorylation in BV-2 microglia from 25-100 µM; induces adiponectin production in 3T3-L1 adipocytes from 2-10 UG/ml; induces the expression of β-cell differentiation genes and increases mRNA levels of Ngn3 and the β-cell markers insulin1 and insulin2 in mPDOs