Learn More
Medchemexpress LLC HY-N4214 5mg Medchemexpress, Isoastragaloside IV CAS:136033-55-1 Purity:>98%
SDP
Catalog No. 501933946
Encompass_Preferred
Medchemexpress, HY-N4214 5mg Isoastragaloside IV CAS:136033-55-1 Isoastragaloside IV is a triterpene oligoglycoside isolated from Astragali Radix. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3946 Supplier Medchemexpress LLC Supplier No. HYN42145MG
Medchemexpress, HY-N4214 5mg Isoastragaloside IV CAS:136033-55-1 Isoastragaloside IV is a triterpene oligoglycoside isolated from Astragali Radix. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.