Learn More
Medchemexpress LLC HY-N0762 5mg Medchemexpress, Isobavachin CAS:31524-62-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN07625MG
Encompass_Preferred
Medchemexpress, HY-N0762 5mg Isobavachin CAS:31524-62-6 Isobavachin, an antioxidant isaolated from Psoralea morisiana with a prenyl group at position 8 of ring A, promotes neuronal differentiation and the potential role of its protein prenylation. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2819