Learn More
Cayman Chemical Isobergapten 10mg
SDP
Supplier:  Cayman Chemical 3414110 MG
Encompass_Preferred
A furanocoumarin with diverse biological activities; inhibits α-glucosidase and scavenges DPPH radicals in cell-free assays (IC50s = 0.385 and 28.6 µM, respectively); active against S. aureus, B. cereus, P. aeruginosa, and S. entritidis (MICs = 125 UG/ml for all); reduces tumor growth in a B16/F10 murine melanoma model at 0.5 mg/kg in combination with UVA radiation
Catalog No. 50-295-0735