Learn More
Alkali Scientific isoBlock™ - Digital Dry Bath, with Two Independently Controlled Chambers, Without Blocks, 115V
SDP
Catalog No. 502336106
Encompass_Preferred
Benchmark’s isoBlock dry bath is ideal for incubation of samples at two different temperatures, in a single compact unit. The two chambers are separated by an insulated wall and are controlled independently, allowing IsoBlock to serve as two dry baths in one. The Hinged (and removable) Lid is also designed with a dividing wall. When closed, this dividing wall acts as a barrier, preventing air transfer between chambers. The control panel features large digital displays of both time and temperature. Both settings of a block can be adjusted at any point, without affecting the settings of the other block. The timer function is non-mandatory; IsoBlock can be used for continuous heating. Various blocks are available for accommodating nearly all popular tube sizes. In addition, the unique Quick-Flip universal blocks accept tubes from 0.5 to 2.0mL and, with just a quick flip to the other side, they also accommodate 0.2mL tubes or PCR strips.
Catalog No. 50-233-6106 Supplier Alkali Scientific Supplier No. ASH6000
Benchmark’s isoBlock dry bath is ideal for incubation of samples at two different temperatures, in a single compact unit. The two chambers are separated by an insulated wall and are controlled independently, allowing IsoBlock to serve as two dry baths in one. The Hinged (and removable) Lid is also designed with a dividing wall. When closed, this dividing wall acts as a barrier, preventing air transfer between chambers. The control panel features large digital displays of both time and temperature. Both settings of a block can be adjusted at any point, without affecting the settings of the other block. The timer function is non-mandatory; IsoBlock can be used for continuous heating. Various blocks are available for accommodating nearly all popular tube sizes. In addition, the unique Quick-Flip universal blocks accept tubes from 0.5 to 2.0mL and, with just a quick flip to the other side, they also accommodate 0.2mL tubes or PCR strips.