Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isoborneol, ≥95.0% (GC), MilliporeSigma™ Supelco™

Catalog No. 111020064 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-102-0064 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-102-0064 Supplier MilliporeSigma Supelco Supplier No. 79856100MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Isoborneol is a monoterpene and a known antioxidant, which is a component of several plant essential oils.

Cleaning Products, Cosmetics, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

CAS 124-76-5
Flash Point 93.4°C - Closed Cup
Linear Formula C10H18 O
MDL Number MFCD00074821
Melting Point 212°C to 214°C (sublimation)
Type Analytical Standard
Quantity 100 mg
Formula Weight 154.25
Percent Purity ≥95.0% (GC)
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.