Learn More
Medchemexpress LLC Isobornyl acetate | 125-12-2 | 50 MG
SDP
Catalog No. 5000420517
Encompass_Preferred
Isobornyl acetate is an aromatic compound used as a food additive for flavoring and as a fragrance in products like perfumes, cosmetics, and detergents. It is for research use only and not sold to patients. Studies indicate certain toxicity to rats at higher doses (15-270 mg/kg), with a no-effect level of 15 mg/kg, which is significantly higher than calculated maximum human intake.
  • Aroma compound
  • Can be used as a food additive for food flavoring
  • Can be used as a fragrance in perfumes, cosmetics, and detergents
Catalog No. 50-004-20517 Supplier Medchemexpress LLC Supplier No. HYN258350MG
Add to Cart
Add to Cart
Isobornyl acetate is an aromatic compound used as a food additive for flavoring and as a fragrance in products like perfumes, cosmetics, and detergents. It is for research use only and not sold to patients. Studies indicate certain toxicity to rats at higher doses (15-270 mg/kg), with a no-effect level of 15 mg/kg, which is significantly higher than calculated maximum human intake.
  • Aroma compound
  • Can be used as a food additive for food flavoring
  • Can be used as a fragrance in perfumes, cosmetics, and detergents