Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Grainger TCI AMERICA Isobornyl Acrylate, Reagent, >90.0% (GC), 25 g

Catalog No. 502780814
Encompass_Preferred

Isobornyl acrylate is a clear, colorless liquid reagent used for laboratory research purposes. This chemical is stabilized with monomethylether hydroquinone (MEHQ) and is provided as a high-purity reagent grade material.

  • Formula: C13H20O2
  • Purity: ≥90.0% (GC)
  • Molecular weight: 208.30
  • Stabilizer: Monomethylether hydroquinone
  • Physical state: Clear, colorless liquid
  • Storage requirements: Store in a cool and dark place

Catalog No. 50-278-0814 Supplier Grainger Supplier No. 19KH37
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Isobornyl acrylate is a clear, colorless liquid reagent used for laboratory research purposes. This chemical is stabilized with monomethylether hydroquinone (MEHQ) and is provided as a high-purity reagent grade material.

  • Formula: C13H20O2
  • Purity: ≥90.0% (GC)
  • Molecular weight: 208.30
  • Stabilizer: Monomethylether hydroquinone
  • Physical state: Clear, colorless liquid
  • Storage requirements: Store in a cool and dark place

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.