Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Isobornyl acrylate

Catalog No. 502891472
Encompass
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-289-1472 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-289-1472 Supplier Sigma Aldrich Supplier No. 3921031L
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Isobornyl acrylate (IBA) can be used to prepare block copolymers with n-butyl acrylate by atom transfer radical polymerization (ATRP).
TRUSTED_SUSTAINABILITY

Specifications

CAS 5888-33-5
Density 0.986 g/mL (at 25°C)
Boiling Point 119°C to 121°C (15 mmHg)
Molecular Formula C13H20O2
Refractive Index n20/D 1.476 (literature)
Linear Formula C13H20O2
Quantity 1 L
MDL Number MFCD00080424
Synonym Acrylic acid isobornyl ester; IBA
Molecular Weight (g/mol) 208.3
Biological Activity Respiratory system
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.