Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isobutene (ca. 8% in Dichloromethane), TCI America™
SDP

Catalog No. I0909100ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I0909100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I0909100ML Supplier TCI America Supplier No. I0909100ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 115-11-7
Molecular Formula C4H8
Molecular Weight (g/mol) 56.11
MDL Number MFCD00008898
InChI Key VQTUBCCKSQIDNK-UHFFFAOYSA-N
Synonym Isobutylene, 2-Methylpropene
PubChem CID 8255
ChEBI CHEBI:43907
IUPAC Name 2-methylprop-1-ene
SMILES CC(C)=C

Specifications

Color Yellow
Quantity 100 mL
UN Number 1593
Formula Weight 56.11
Physical Form Liquid
Chemical Name or Material Isobutene (ca. 8% in Dichloromethane)
EINECSNumber (2)-0016
RTECSNumber UD0890000
TSCA Yes
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.