Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Isobutyl 1,2-Dihydro-2-Isobutoxy-1-Quinolinecarboxylate

Catalog No. 17824125G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
17-824-125G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 17-824-125G Supplier Sigma Aldrich Supplier No. 17824125G
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Isobutyl 1,2-Dihydro-2-Isobutoxy-1-Quinolinecarboxylate

Specifications

CAS 144789-75-3
Molecular Formula C9H8F3NO2
Refractive Index n20/D 1.523
Linear Formula C9H8F3NO2
MDL Number MFCD00236329
Quantity 1 g
Synonym 1-Isobutoxycarbonyl-2-isobutoxy-1,2-dihydroquinoline; 2-Isobutoxy-1-isobutoxycarbonyl-1,2-dihydroquinoline; IIDQ
Molecular Weight (g/mol) 219.16 g/mol
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.