Learn More
Crescent Chemical Co Inc ISOBUTYL-1-METHYLXANTHINE
SDP
Catalog No. 50247752
Encompass_Preferred
Isobutyl-1-methylxanthine research grade Inhibitor of cAMP-phosphodiesterase. Soluble in ethanol. Size - 500MG Storage Conditions - +15 °C TO +30 °C Catalog Number - 26445.02 CAS 28822-58-4
Catalog No. 50-247-752 Supplier Crescent Chemical Co Inc Supplier No. SE26445.02
Isobutyl-1-methylxanthine research grade Inhibitor of cAMP-phosphodiesterase. Soluble in ethanol. Size - 500MG Storage Conditions - +15 °C TO +30 °C Catalog Number - 26445.02 CAS 28822-58-4