Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Isobutyl 4-hydroxybenzoate | 4247-02-3 | MFCD00020167 | 98.9% | 194.23 g·mol⁻¹ | C11H14O3 | 25 G
SDP

Catalog No. 5000303475
Encompass_Preferred

Isobutylparaben (isobutyl 4-hydroxybenzoate) is a research-grade paraben used as an antimicrobial preservative and biochemical reagent. It is supplied as a white to off-white solid with high purity for biological and analytical studies, including receptor activity and toxicology research.

  • High purity: 98.9% (as supplied).
  • Agonist of PXR, CAR, and PPAR for nuclear receptor studies.
  • Broad-spectrum antimicrobial properties; used as a preservative model.
  • Chemical identifiers: CAS 4247-02-3; formula C11H14O3; MW 194.23 g·mol⁻¹.
  • Suitable for biochemical, analytical, and toxicology research applications.

Catalog No. 50-003-03475 Supplier Medchemexpress LLC Supplier No. 25GHYW015026
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Isobutylparaben (isobutyl 4-hydroxybenzoate) is a research-grade paraben used as an antimicrobial preservative and biochemical reagent. It is supplied as a white to off-white solid with high purity for biological and analytical studies, including receptor activity and toxicology research.

  • High purity: 98.9% (as supplied).
  • Agonist of PXR, CAR, and PPAR for nuclear receptor studies.
  • Broad-spectrum antimicrobial properties; used as a preservative model.
  • Chemical identifiers: CAS 4247-02-3; formula C11H14O3; MW 194.23 g·mol⁻¹.
  • Suitable for biochemical, analytical, and toxicology research applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.