Learn More
Medchemexpress LLC Isobutyl 4-hydroxybenzoate | 4247-02-3 | MFCD00020167 | 98.9% | 194.23 g·mol⁻¹ | C11H14O3 | 500 G
SDP
Catalog No. 5000303169
Encompass_Preferred
Isobutyl 4-hydroxybenzoate (isobutylparaben) is a paraben-class preservative supplied as a solid research reagent. It is used in biochemical studies as a receptor agonist and in formulation research for its antimicrobial properties and compatibility with common laboratory solvents and in vivo vehicles.
  • High purity suitable for analytical and research applications.
  • Broad-spectrum antimicrobial activity useful in formulation studies.
  • Characterized molecular formula C11H14O3 and molecular weight 194.23 g·mol⁻¹.
  • Soluble in DMSO; compatible with common in vivo formulation vehicles at tested concentrations.
  • Stable as a solid under recommended storage conditions.
Catalog No. 50-003-03169 Supplier Medchemexpress LLC Supplier No. HYW015026500G
Isobutyl 4-hydroxybenzoate (isobutylparaben) is a paraben-class preservative supplied as a solid research reagent. It is used in biochemical studies as a receptor agonist and in formulation research for its antimicrobial properties and compatibility with common laboratory solvents and in vivo vehicles.
  • High purity suitable for analytical and research applications.
  • Broad-spectrum antimicrobial activity useful in formulation studies.
  • Characterized molecular formula C11H14O3 and molecular weight 194.23 g·mol⁻¹.
  • Soluble in DMSO; compatible with common in vivo formulation vehicles at tested concentrations.
  • Stable as a solid under recommended storage conditions.