Learn More
Medchemexpress LLC Isobutyl 4-hydroxybenzoate-d4 | 1219805-33-0 | 98.5% | 198.25 g/mol | C11H10D4O3 | 10 MG
SDP
Catalog No. 5000303166
Encompass_Preferred
Isobutylparaben-d4 is the deuterium-labeled analogue of isobutylparaben used as an internal standard and analytical reference for LC-MS and related assays. It is provided as a solid at high purity for quantitative analysis and method development.
  • Deuterium-labeled internal standard for analytical assays.
  • High purity suitable for quantitative LC-MS applications.
  • Available in small vial sizes for laboratory use.
  • Stable isotope provides accurate quantification and traceability.
  • Compatible with common organic solvents and sample preparation workflows.
Catalog No. 50-003-03166 Supplier Medchemexpress LLC Supplier No. HYW015026S10MG
Isobutylparaben-d4 is the deuterium-labeled analogue of isobutylparaben used as an internal standard and analytical reference for LC-MS and related assays. It is provided as a solid at high purity for quantitative analysis and method development.
  • Deuterium-labeled internal standard for analytical assays.
  • High purity suitable for quantitative LC-MS applications.
  • Available in small vial sizes for laboratory use.
  • Stable isotope provides accurate quantification and traceability.
  • Compatible with common organic solvents and sample preparation workflows.