Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isobutyl acetate, Analytical Standard, >99.8% (GC), MilliporeSigma™ Supelco™

Catalog No. 111020279 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-102-0279 1 mL
11-101-8804 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-102-0279 Supplier MilliporeSigma Supelco Supplier No. 948231MLF
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Isobutyl acetate is a chemical compound, commonly used as a solvent and flavoring agent in food products.

TRUSTED_SUSTAINABILITY

Specifications

CAS 110-19-0
Vapor Pressure 15 mmHg (20°C)
Vapor Density >4 (vs. air)
Density 0.867 g/mL (at 25°C)
Physical Form Neat
Linear Formula CH3COOCH2CH(CH3)2
MDL Number MFCD00008932
Melting Point -99°C
Boiling Point 115°C to 117°C
Synonym Acetic acid isobutyl ester
Quantity 1 mL
Formula Weight 116.16
Percent Purity >99.8% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.