Learn More
Sigma Aldrich Fine Chemicals Biosciences Isobutyl acetate fCC fG
Catalog No. 502903915
Encompass
Isobutyl acetate a medium chain ester is a high-volume solvent flavor and fragrance compound. It is the fruity ester formed during the fermentation of wine. Organoleptic threshold value of isobutyl acetate has been evaluated in Norwegian all-malt pilsener beer. Engineered biosynthesis of isobutyl acetate in Escherichia coli directly from glucose has been reported.
Catalog No. 50-290-3915 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. W21750620KGK
Isobutyl acetate a medium chain ester is a high-volume solvent flavor and fragrance compound. It is the fruity ester formed during the fermentation of wine. Organoleptic threshold value of isobutyl acetate has been evaluated in Norwegian all-malt pilsener beer. Engineered biosynthesis of isobutyl acetate in Escherichia coli directly from glucose has been reported.